Micro-Spatula
Spatula, Micro, Stainless Steel, 21 cm (8.3")
Catalog # 470008-074
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: SSFT08
Jump to:
Product Details & Documents
This spatula is great for handling small amount of chemicals.
- Stainless steel construction
- One pointed end
This micro spatula is perfect for mixing and dispensing small volume of powders or solutions.
Especificaciones
- Description:Spatula, Micro, Stainless Steel
- Length:21 cm (8.3")