Microcentrifuge Tube Rack
Catalog # 470100-896
Supplier: PRECISION MOULDED POLYMERS LTD
Specifications
- Descripción:Microcentrifuge Tube Racks
- Dimensiones:15×13×2.5 cm
- Referencia:470100-896
- Nº de proveedor:A-M9001
Specifications
About this item
Microcentrifuge tube racks are clear acrylic tube rack holds 25 standard 1.5 ml microcentrifuge tubes.
Packaging: Package of 10.