Orders LinkContactUsLinkComponent
Metallic Mineral Resources Topic Set
Metallic Mineral Resources Topic Set
Metallic Mineral Resources Topic Set, 12
Catalog # 470025-360
Supplier:   Avantor
Metallic Mineral Resources Topic Set
Catalog # 470025-360
Supplier:  Avantor
Metallic Mineral Resources Topic Set
Metallic Mineral Resources Topic Set
Metallic Mineral Resources Topic Set, 12
Price
$1,433.40
Log in Para disponibilidad
Kit de 1
Product Details & Documents
Give Your Students a Lesson in Economics as Well as Geology
  • Developed by teachers and WARD's Staff Geologists
  • Classroom tested for dependabilty
  • Designed to save valuble planning and preparation time
Students will learn the economic importance of metallic minerals with this comprehensive GEO-logic set. A teacher's guide and student copymasters are included for the following activities: grouping according to type, household uses of minerals, and worldwide distribution of metals. The set is housed in a compartmented collection box.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides. The types of GEO-logic teaching collections include:
Topic Sets, which provide greater depth in an individual subject area and include both a teacher’s guide and student copymasters
Classroom Bundles, which provide 15 bulk-packaged samples included in the reference or topic set and 15 sets of any required testing materials included with the corresponding set
*Classroom Bundles are bulk-packaged and do not include display boxes, or teacher/student activity guides.
Contents:12 numbered samples, 1" x 2"
Minerals:bauxite, beryl, cassiterite, chromite, galena, hematite, ilmenite, molybdenite, native copper, psilomelane, smaltite, sphalerite
Especificaciones
  • Description:
    Metallic Mineral Resources Topic Set
  • No. of Samples:
    12