SP Bel-Art Mercury Collector, Bel-Art Products, a part of SP
470153-828
- Descripción:Mercury Collector
- Referencia:470153-828
- Nº de proveedor:F362500000
Simply press the foam collector onto elusive beads of spilled mercury and the foam pores open to enclose it. Closing the container forces the beads into a recessed trap. One replacement pad is included.