Orders LinkContactUsLinkComponent
Medium Grade, 15 μm
Medium Grade, 15 μm
Certificates
Medium Grade, 15 μm
Medium Grade, 15 μm
4 Options Available Below
Product Details & Documents
The most widely used qualitative filter paper for routine applications.
  • Medium grade, 15 μm
Holds flocculants and precipitates commonly encountered in laboratory exercises.
 : Packages of 100.
Product Family Options
Información del producto
  • Availability
    Price
  • 470019-882
    Filter Paper, Medium Grade, 90 mm diameter
    Filter Paper, Medium Grade, 90 mm diameter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Caja de 100
    Caja de 100 - $55.96
    $55.96
    Caja de 100
       $55.96
    Supplier Number: FPR009
  • 470019-884
    Filter Paper, Medium Grade, 110 mm diameter
    Filter Paper, Medium Grade, 110 mm diameter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Caja de 100
    Caja de 100 - $192.95
    $192.95
    Liquidación
    Caja de 100
       $192.95
    Supplier Number: FPR011
  • 470019-886
    Filter Paper, Medium Grade, 125 mm diameter
    Filter Paper, Medium Grade, 125 mm diameter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Caja de 1
    Caja de 1 - $112.47
    $112.47
    Caja de 1
       $112.47
    Supplier Number: FPR125
  • 470019-888
    Filter Paper, Medium Grade, 150 mm diameter
    Filter Paper, Medium Grade, 150 mm diameter
    Log in Para disponibilidad
    Log in Para disponibilidad
    Caja de 1
    Caja de 1 - $157.77
    $157.77
    Caja de 1
       $157.77
    Supplier Number: FPR150