Aneroid Sphygmomanometer
Aneroid Sphygmomanometer
Catalog # 470019-064
Supplier: HEALTH SMART SE
Número de proveedor #: 01-100-011
Jump to:
Product Details & Documents
Features Markings On The Cuff For Better Accuracy
Pre-calibrated and self-zeroing, this model assures you consistently accurate measurements. It comes with a standard-size cuff with Velcro closure, instructions, and stores in its own leatherette case. Accurate to ±3 mm Hg.
Especificaciones
- Description:Aneroid Sphygmomanometer