Orders LinkContactUsLinkComponent
Spoon Set
Spoon Set
Set of 6 Measuring Spoons: 1/8 tsp (0.62 mL), 1/4 tsp (1.25 mL), 1/2 tsp (2.5 mL), 1 tsp (5 mL), 1/2 tbsp. (7.5 mL), and 1 tbsp. (15 mL)
Catalog # 470016-906
Supplier #:  12012
Spoon Set
Catalog # 470016-906
Spoon Set
Spoon Set
Set of 6 Measuring Spoons: 1/8 tsp (0.62 mL), 1/4 tsp (1.25 mL), 1/2 tsp (2.5 mL), 1 tsp (5 mL), 1/2 tbsp. (7.5 mL), and 1 tbsp. (15 mL)
Price
$43.16
Log in for availability
Kit de 5
Product Details & Documents
Teach Students About Measurements with these Handy Sets
  • Made with sturdy polypropylene
  • Measurement marked on handle
  • Grades K+
  • Set includes 1 each of 15 ml, 7.5 ml, 5 ml, 2.5 ml, 1.25 ml and 0.625 ml (1 tbsp, 1 1/2, 1, 1/2, 1/4 and 1/8 tsp)
Specifications
  • Description:
    Set of 6 Measuring Spoons: 1/8 tsp (0.62 mL), 1/4 tsp (1.25 mL), 1/2 tsp (2.5 mL), 1 tsp (5 mL), 1/2 tbsp. (7.5 mL), and 1 tbsp. (15 mL)