Measuring Experiments Student Activity
Measuring Experiments Student Activity, Teacher's Guide, Student Guides and Worksheets, SDS, 12
470106-538
: 102
Science Students at Any Level Must Understand Various Units of Measurement and How to Use Them
- Measurement Concepts Include:
- Measure the distance to the Sun
- Consider magnitude of the Solar System
- Determine "size" of low moon vs. high moon
- Determine diameter of the moon
- And more!
- Requires one 50 minutes classroom periods to complete
Students need to scientific measurement, particularly with respect to magnitude and computation. In this kit students perform 12 exercises covering measuring common objects; utilizing information derived from graphs; determining the Earth’s radius, diameter and circumference; and the use of baselines and a range-finder to determine distance.
: Materials include Brass clips, LAB-AIDS® rangefinder/earth rules, Magnets, Marbles, String, Paper Strips, Protractors, Graph paper, Straws, Weights, Sets of Student Activity Cards #1-#4.
- Description:Measuring Experiments Student Activity
- No. of Lab Groups:12
- Includes:Teacher's Guide, Student Guides and Worksheets, SDS