Sand and Gravel
470103-690
Small Business Enterprise
- Descripción:Gravel, Aquarium, Double Bagged
- Referencia:470103-690
- Nº de proveedor:6024005
Sand, pebbles, and gravel are all ideal for earth science experiments and simulated excavations.
Weight: 2.5 kg (5.5 lbs.)