Mast Cells Slide
Mast Cells, Whole mount, Fuchsin and Hematoxylin
Catalog # 470182-634
Supplier: Avantor
Jump to:
Product Details & Documents
Flat Mount of Mesentery
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Resorcin fuchsin stained to demonstrate mast cells as well as both white and elastic fibers.
Specifications
- Description:Mast Cells
- Stain:Fuchsin and Hematoxylin
- Orientation:Whole mount