Mass Hangers
Supplier: STEINER ENTERPRISES SE
Jump to:
Acerca de este artículo
Replacement hanger for slotted masses.
A top hook and precise weight make this stainless steel hanger ideal for use with slotted weights
2 Options Available Below
Product Details & Documents
Replacement hanger for slotted masses.
A top hook and precise weight make this stainless steel hanger ideal for use with slotted weights
Product Family Options
Información del producto
- WeightAvailabilityPrice
- 470016-120Mass Hanger, 500 g Capacity
- 470017-096Mass Hanger, 1500 g Capacity