Markable Microscope Slides
Markable slides, package of 72, 75×25 mm (3×1")
Catalog # 470019-018
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: MSLF01
Jump to:
Product Details & Documents
Frosted Labeling Area Allows for Easy Reading
- Quality slide making supplies
- Fast and easy labeling on frosted surface
There’s no need for sticky slide labels that could slip or fall off because these optically clear slides have a frosted coating on one edge that’s easy to mark and read.
Especificaciones
- Description:Markable slides, package of 72
- L×W:75×25 mm (3×1")