Orders LinkContactUsLinkComponent
Ward's® Marble (Fine White)
Ward's® Marble (Fine White)
Supplier:   Avantor
Ward's® Marble (Fine White)
Supplier:  Avantor
Certificates
Ward's® Marble (Fine White)
Ward's® Marble (Fine White)
3 Options Available Below
Product Details & Documents
Fine crystalline, pure calcite.
  • Collected in Vermont
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Specimen Dimensions
    Availability
    Price
  • 470015-482
    Marble (Fine White), student specimens
    Marble (Fine White), student specimens
    1×1 - 1×1"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $229.68
    $229.68
    Paquete de 10
       $229.68
  • 470015-484
    Marble (Fine White), hand specimen
    Marble (Fine White), hand specimen
    2×3" - 3×4"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $293.25
    $293.25
    Each (1UN)
       $293.25
  • 470026-112
    Marble (Fine White), bulk pack, 1 kg
    Marble (Fine White), bulk pack, 1 kg
    Size will vary
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $343.65
    $343.65
    Each (1UN)
       $343.65