Orders LinkContactUsLinkComponent
Ward's® Magnetite (Lodestone)
Ward's® Magnetite (Lodestone)
Supplier:   Avantor
Ward's® Magnetite (Lodestone)
Supplier:  Avantor
Certificates
Ward's® Magnetite (Lodestone)
Ward's® Magnetite (Lodestone)
3 Options Available Below
Product Details & Documents
Spinel group; iron-black, massive; natural magnet.
  • Collected in Utah
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Specimen Dimensions
    Availability
    Price
  • 470015-432
    Magnetite (Lodestone), Student Specimens
    Magnetite (Lodestone), Student Specimens
    1×1 - 1×1"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $849.35
    $849.35
    Paquete de 10
       $849.35
  • 470015-434
    Magnetite (Lodestone), Hand Specimen, Each
    Magnetite (Lodestone), Hand Specimen, Each
    2×3" - 3×4"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $663.60
    $663.60
    Each (1UN)
       $663.60
  • 470025-678
    Magnetite (Lodestone), Bulk Pack, 1 kg
    Magnetite (Lodestone), Bulk Pack, 1 kg
    Size will vary
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $655.20
    $655.20
    Each (1UN)
       $655.20