Magnetic Field Demonstrator
Magnetic Field Demonstrator
Catalog # 470002-988
Supplier: VWR International
Jump to:
Product Details & Documents
Convenient and Easy to Use
- Includes a powerful magnet
Reveal the magnetic lines of forces when 48 compasses are fixed around a magnet. This simple demonstration visually realizes magnetic fields and is an ideal introduction to magnetics.
Specifications
- Description:Magnetic Field Demonstrator