Magnetic Compass with Beveled Top
470001-824
- Descripción:Magnetic compass with beveled top
- Referencia:470001-824
- Nº de proveedor:CWR045-W
This simple, student-grade model is not dampened and measures in 2° increments.
- It is brass-colored and has a ring for attaching a lanyard
- Great price makes it ideal for outfitting large classes
Size: 2 Ø.×1/2"