Magna Atom Magnetic Chalkboard Model Set
Catalog # 470004-744
Supplier: KELLER BROS. & MILLER, INC.
Specifications
- Descripción:Magna Atom Magnetic Chalkboard Model Set
- Referencia:470004-744
- Nº de proveedor:4738800
Specifications
About this item
Demonstrate Relationships Between Atoms
- Whole Class Hands-On Demonstration
- Rearrange subatomic particles to form new atoms/isotopes
Students can easily rearrange the electrons, protons, and neutrons to form different atoms or isotopes. Then, they can identify the atomic numbers of elements, distinguish between various elemental isotopes, or compare covalent and ionic bonding. It comes with enough materials to simulate elements and their isotopes up to atomic number 24.