Orders LinkContactUsLinkComponent
Magic Garden Kit
Magic Garden Kit
Kit, Crystal Growing, Magic Garden
Catalog # 470004-442
Supplier:   SCHYLLING, INC
Número de proveedor #:  MG
Magic Garden Kit
Catalog # 470004-442
Supplier:  SCHYLLING, INC
Magic Garden Kit
Magic Garden Kit
Kit, Crystal Growing, Magic Garden
Price
$159.92
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Grow your own beautiful mountain scene with this kit's mountain, trees and flowers made of paper and a plastic base.
  • Once you've assembled your paper garden, pour the magic garden solution on the paper
  • Within hours, the flowers bloom, the trees bud and snow appears on the mountain
This spectacular scene and its metamorphosis will entertain and educate about crystallization creation and formation.
Caution: WARNING: CHOKING HAZARD- Small parts not for children under 3 years.
Especificaciones
  • Description:
    Kit, Crystal Growing, Magic Garden
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 