Lymphatic Vessel Slide
Lymphatic Vessel, Cross section, Hematoxylin and Eosin
Catalog # 470182-770
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates a Component of the Lymphatic System
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Carries lymph throughout the body.
Especificaciones
- Description:Lymphatic Vessel
- Stain:Hematoxylin and Eosin
- Orientation:Cross section