Lung Slide
Lung Slide, Section, Mallory Triple
470182-904
: Avantor
Lung characteristics illustrated
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal lobule. Shows alveolar sacs and small bronchioles.
- Description:Lung Slide
- Stain:Mallory Triple
- Orientation:Section