Lung Slide
Lung Slide, Section, Hematoxylin, phloxine and saffron
470182-906
: Avantor
Specific structures of the lung illustrated
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal lobule. Shows alveoli as well as respiratory bronchioles and bronchi.
- Description:Lung Slide
- Stain:Hematoxylin, phloxine and saffron
- Orientation:Section