Lung Slide
Lung, Section, Hematoxylin and orcein
470182-910
: Avantor
Illustrates Lung Structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal lobule. Shows elastic fibers in the lamina propria.
- Description:Lung
- Stain:Hematoxylin and orcein
- Orientation:Section