Lunar Cycle Banner
Lunar Cycle Banner, 48", 12"
Catalog # 470016-828
Supplier: BRITTEN BANNER INC.
Supplier #: 7396101
Jump to:
Product Details & Documents
Sized for Viewing Throughout the Class
- Displays names of eight phases
A series of photographic images showing the changing face of the moon over a 28-day period, this stellar visual aid features the names of eight phases and an indication of waxing or waning moons. Poster is made from poly vinyl stock with three grommets across the top for hanging. Size: 48" x 12".
Specifications
- Description:Lunar Cycle Banner
- Length:48"
- Width:12"