Orders LinkContactUsLinkComponent
Refraction Blocks
Refraction Blocks
Refraction Blocks
Refraction Blocks
Rectangular blocks in a variety of sizes and are designed for use as an index of refraction plate.
  • Quality Lucite block
  • Optically clear sides
This Lucite block allows you to investigate refraction and internal reflection.
Product Family Options
  • Description
    Dimensions
  • Refraction Block
    75 x 50 x 15 mm
    Each (1UN)
    Each (1UN) - $219.61
    $219.61
    Each (1UN)
       $219.61
     : RCB075
  • Refraction Block
    100 x 75 x 20 mm
    Each (1UN)
    Each (1UN) - $252.66
    $252.66
    Each (1UN)
       $252.66
     : RCB100