Lets Read and Find out Science
Bugs Are Insects, (Rockwell), 32, 19 cm, 23 cm
Catalog # 470047-774
Supplier: HOUGHTON MIFFLIN HARCOURT (HMH) SE
Número de proveedor #: 9780062381828
Jump to:
Product Details & Documents
Beginners science readings addressing common questions new readers would have.
Learn how seeds grow, what makes you sick, why leaves change color, and other interesting topics in these beautifully illustrated books.
Especificaciones
- Description:Bugs Are Insects, (Rockwell)
- Length:19 cm
- No. of Pages:32
- Width:23 cm