Low Stender-Form Preparation Dish
Low Stender-Form Preparation Dish
Catalog # 470144-866
Supplier: VARIETY GLASS INC
Supplier #: 679
Jump to:
Product Details & Documents
Dish is made of clear pressed glass. Cover is made of plain pressed glass and ground to fit. Size: 60mm Dia. x 29mm H.
Made of flint glass. Must be heated and cooled slowly. Can be heated up to 285 degrees Celsius.
Specifications
- Description:Low Stender-Form Preparation Dish