Economy Alcohol Burner Stand
470104-982
- Descripción:Burner Stand
- Altura:11 cm
- Referencia:470104-982
- Nº de proveedor:ABSP05
This rugged wire support stand has a heavy wire gauze top.
- Wire gauze surface measures 8×10 cm (3×2")
- Height: 11.5 cm (4½")
Made especially for alcohol burners.