Long Foam Tipped Brush
Foam Tipped Long Laboratory Glassware Washing Brush, Foam, 20"
Catalog # 470017-821
Supplier: BRUSHTECH
Supplier #: B208C
Jump to:
Product Details & Documents
Features a wide flat handle that is easy to rotate and manuever, ideal to clean labware.
- Ergonomically designed
- No detergent is needed
- Length: 20" including handle
For use with both glass and plastic, the brush effectively cleans wide-mouthed lab bottles, culture flasks, volumetric and filtering flasks, jugs, and much more.
Especificaciones
- Bristle Material:Foam
- Description:Foam Tipped Long Laboratory Glassware Washing Brush
- Total length:20"