Orders LinkContactUsLinkComponent
Long Foam Tipped Brush
Long Foam Tipped Brush
Foam Tipped Long Laboratory Glassware Washing Brush, Foam, 20"
Catalog # 470017-821
Supplier:   BRUSHTECH
Supplier #:  B208C
Long Foam Tipped Brush
Catalog # 470017-821
Supplier:  BRUSHTECH
Long Foam Tipped Brush
Long Foam Tipped Brush
Foam Tipped Long Laboratory Glassware Washing Brush, Foam, 20"
Price
$204.24
Log in Para disponibilidad
Each (1UN) Case de 48 (1UN)
Product Details & Documents
Features a wide flat handle that is easy to rotate and manuever, ideal to clean labware.
  • Ergonomically designed
  • No detergent is needed
  • Length: 20" including handle
For use with both glass and plastic, the brush effectively cleans wide-mouthed lab bottles, culture flasks, volumetric and filtering flasks, jugs, and much more.
Especificaciones
  • Bristle Material:
    Foam
  • Description:
    Foam Tipped Long Laboratory Glassware Washing Brush
  • Total length:
    20"