Liver Slide
Liver Slide, Section, Mallory Triple
Catalog # 470182-884
Supplier: Avantor
Jump to:
Product Details & Documents
Demonstrates pig-specific liver characteristics
- Identifying features clearly distinguishable
- Rigorous quality control standards
Pig. Demonstrates the lobular structure of the liver through heavily developed connective tissue, a peculiarity of pig liver.
Especificaciones
- Description:Liver Slide
- Stain:Mallory Triple
- Orientation:Section