Orders LinkContactUsLinkComponent
Ward's® Limestone (Gray)
Ward's® Limestone (Gray)
Supplier:   Avantor
Ward's® Limestone (Gray)
Supplier:  Avantor
Certificates
Ward's® Limestone (Gray)
Ward's® Limestone (Gray)
3 Options Available Below
Product Details & Documents
Fine, common limestone; massive, and will fizzle with HCl.
  • Collected in New York
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Specimen Dimensions
    Availability
    Price
  • 470103-720
    Limestone (Gray), Student Specimens
    Limestone (Gray), Student Specimens
    1×1 - 1×1"
    Log in for availability
    Log in for availability
    Paquete de 10
    Paquete de 10 - $214.78
    $214.78
    Paquete de 10
       $214.78
  • 470103-722
    Limestone (Gray), Hand Specimen
    Limestone (Gray), Hand Specimen
    2×2" - 3×4"
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $236.55
    $236.55
    Each (1UN)
       $236.55
  • 470026-058
    Limestone (Gray), Bulk Pack, 1 kg
    Limestone (Gray), Bulk Pack, 1 kg
    Size will vary
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $289.80
    $289.80
    Each (1UN)
       $289.80