Lil'pet Micropipette
Lil'pet Micropipettes feature ideal volumes for gel loading.
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
Lil'pet Micropipettes feature ideal volumes for gel loading.
- Units accept standard micropipette tips
- Economical
- Durable
- Autoclavable
These fixed-volume pipettes feature a double stop plunger to expel all liquid from the tip.
The 1000 μl model includes two tips. All other pipettes come with one each.
Product Family Options
- Volume
- 5 µlEach (1UN)$279.06Case de 100 (1UN)$304.18
- 10 µlEach (1UN)$279.06Case de 100 (1UN)$304.18
- 20 µlEach (1UN)$6.70Case de 100 (1UN)$670.00
- 50 µlEach (1UN)$279.06Case de 100 (1UN)$304.18
- 100 µlEach (1UN)$279.06Case de 100 (1UN)$304.18
- 250 µlEach (1UN)$345.18Case de 100 (1UN)$376.25
- 500 µlEach (1UN)$345.18Case de 100 (1UN)$376.25
- 1000 µlEach (1UN)$279.06Case de 100 (1UN)$304.18