Lightning® Gel Tape
Lightening Gel Tape
Catalog # 470019-982
Supplier: FORENSICS SOURCE
Número de proveedor #: 1004131
Jump to:
Product Details & Documents
Ideal for Use on Textured Surfaces
- Apply the tape initially as you would normal lifting tape, but expect extraordinarily detailed prints.
Featuring a thick, pliable padding that is over 20 times as thick as standard lifting tapes, this fingerprint tape is ideal for lifting details from uneven and textured surfaces. Apply the tape initially as you would normal lifting tape, but expect extraordinarily detailed prints. After application, press the gel tape down into the grooves of the surface. Then, simply lift the tape and place the lifted print onto a backing card for analysis and examination.
Especificaciones
- Description:Lightening Gel Tape