Light Source
Light Source, Replacement Bulb
Catalog # 470111-326
Supplier: NORMAN LAMPS INC SE
: 7.5S11-13VFR
Jump to:
Product Details & Documents
A handy addition to your optical bench, this light source features a 7.5 watt incandescent lamp plus a cord and plug for connection to 110 VAC power source.
Especificaciones
- Description:Light Source, Replacement Bulb