Orders LinkContactUsLinkComponent
Rudiger® Life-Size Human Skeleton Model
Rudiger® Life-Size Human Skeleton Model
Rudiger® Life-Size Human Skeleton Model
Certificates
Rudiger® Life-Size Human Skeleton Model
Rudiger® Life-Size Human Skeleton Model
2 Options Available Below
Product Details & Documents
These high-quality, synthetic skeletons are perfect for everyday use.
  • Five-Year Manufacturer's Warranty
  • Durable Plastic Molding
  • Excellent Articulation
  • Adjustable Plastic Stand Included
  • Vinyl Dust Cover Included
Each skeleton is an accurate, life-size replica of an adult, human male and features horizontal sectioning of the skull to permit study of the cranial cavity. They include an adjustable plastic stand and a zippered, vinyl dust cover.
The model with painted muscles has muscle origins painted in red and insertions painted in blue on its left side. Each muscle is numbered for easy identification. The right side of this skeleton has been left unpainted for bone study.
Ordering information: This product includes a manufacturer's warranty.
Product Family Options
Información del producto
  • No. of Parts
    Availability
    Price
  • 470007-324
    Skeleton
    Skeleton
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $13,777.71
    $13,777.71
    Each (1UN)
       $13,777.71
    Supplier Number: A200
  • 470106-080
    Painted Skeleton
    Painted Skeleton
    1
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $24,966.60
    $24,966.60
    Each (1UN)
       $24,966.60
    Supplier Number: A200.1