Orders LinkContactUsLinkComponent
Lens Sets
Lens Sets
Lens Sets
Lens Sets
Choose high quality crown glass or shatter-resistant acrylic lenses.
  • Wide range of lens sizes and materials
  • 6 lenses: double convex, plano-convex, concavo-convex, double concave, plano-concave, and convexo-concave
Perfect for optical benches or general use, these sets contain one each of the six lens types shown. Glass lenses resist scratching, but are more brittle than acrylic. Sets come in a cardboard storage box. Acrylic sets include an activity guide. Available in 38 mm or 50 mm diameter.
Product Family Options
  • Description
    Diameter
    Material
  • Lens Set
    38 mm
    Glass
    Kit de 6
    Kit de 6 - $157.24
    $157.24
    Kit de 6
       $157.24
  • Lens Set
    50 mm
    Glass
    Kit de 6
    Kit de 6 - $173.22
    $173.22
    Kit de 6
       $173.22
  • Lens Set
    50 mm
    Acrylic
    Each (1UN)
    Each (1UN) - $8.00
    $8.00
    Each (1UN)
       $8.00