Orders LinkContactUsLinkComponent
Lens Paper
Lens Paper
Lens Paper
Lens Paper
This soft, absorbent paper cleans lenses and optical elements.
  • Available in booklet and sheet form
The paper removes grit, lint, and dirt from cleaning lenses and optical elements.
Product Family Options
  • Description
    Length
    Width
  • Lens Paper, Form: Loose Sheets, Sheets
    12"
    9"
    Paquete de 50
    Paquete de 50 - $140.20
    $140.20
    Paquete de 50
       $140.20
     : 20 20 5103
  • Lens Paper, Form: Booklet, Sheets
    6"
    4"
    Each (1UN)
    Each (1UN) - $33.88
    $33.88
    Case de 12 (1UN)
    Case de 12 (1UN) - $36.93
    $36.93
    Each (1UN)
       $33.88
    Case de 12 (1UN)
       $36.93
     : 20205118