Lens and Prism Set
470013-300
Small Business Enterprise
- Descripción:Lens and Prism Set, Acrylic
- Referencia:470013-300
- Nº de proveedor:3358-20
A Staple For Optic Studies
These sets contain seven different lenses and prisms to demonstrate a wide range of optical principles. The frosted surfaces are also excellent for revealing the path of light as it passes through the lens.