Leaf Pack Macroinvertebrate Flash Cards
Macroinvertebrate Flash Cards, 18 Cards, 15.2 cm, 20 cm
Catalog # 470015-846
Supplier: LA MOTTE CHEMICAL PROD CO SE
Supplier #: 5882-SA1
Jump to:
Product Details & Documents
Strong Presentation of Insects and Other Major Classes
- Handy, durable flashcards
Each card features a vivid, close-up photograph of a living invertebrate on the front. The back of the card features detailed form and structure information for the invertebrate, including classification, key characteristics, illustration of the specimen, and specimen size scale.
Specifications
- Description:Macroinvertebrate Flash Cards, 18 Cards
- Length:15.2 cm
- Width:20 cm