Lattice Clamp
1.3 cm (0.5")
Catalog # 470123-514
Supplier: VWR International
Jump to:
Product Details & Documents
This clamp is used for lattice building and rod coupling.
- Clamp Fitted with Allen Wrench Screws
- Allen Wrench Included
- For Lattice Building and Rod Coupling
This lattice clamp is fitted with screws that match Allen Wrenches.
An Allen Wrench is included.
Ordering information: This clamp includes one Allen Wrench.
Especificaciones
- Rod Diameter:1.3 cm (0.5")