Latex-Free Disposable Gloves
XL, Translucent, 22.8 cm (9"), Box/100
Catalog # 470150-920
Supplier: DIVAL SAFETY & SUPPLIES SE
Supplier #: GVDRXLIC
Jump to:
Product Details & Documents
Convenient disposable gloves.
- Powdered
- Ambidextrous
- Nonsterile
These powdered vinyl gloves provide the same fit and protection as latex gloves, without the irritation that affects some people.
Specifications
- Glove Color:Translucent
- Glove size:XL
- Length:22.8 cm (9")
- Packaging:Box/100
- Cat. No.:470150-920
- Latex / latex-free:Latex-free
- Disposable / reusable:Disposable
- Powdered / powder-free:Powdered
- Ambidextrous / hand-specific:Ambidextrous