Latent Fingerprinting Supplies
Fingerprint Brush, Camel Hair, 6¼"
Catalog # 470149-656
Supplier: GORDON BRUSH MFG CO
Supplier #: 0187-00001
Jump to:
Product Details & Documents
Develop and study otherwise scarcely visible fingerprints using high-quality fingerprint brushes.
- Different fingerprinting brushes to suit your preferences
- Supplies Demonstrate the minute differences in individual fingerprints
- Camel Brush Ideal for General Conditions
- Zephyr Brush Retains a Large Amount of Powder
- Brush Length: 61/4"
For general conditions, the camel hair brush is ideal.
The Zephyr fiberglass brush retains a larger amount of powder and applies it more evenly to provide clear prints in any circumstance.
Especificaciones
- Description:Fingerprint Brush, Camel Hair
- Length:6¼"