Orders LinkContactUsLinkComponent
Dissecting Pans And Sets
Dissecting Pans And Sets
Supplier:   PAOLI CLAY CO SE
Dissecting Pans And Sets
Supplier:  PAOLI CLAY CO SE
Certificates
Dissecting Pans And Sets
Dissecting Pans And Sets
6 Options Available Below
Product Details & Documents
The provided aluminum pans resist corrosion.
  • Aluminum Pans Resist Corrosion
  • Set Sizes between 11 and 30 Inches
The standard-size set is about 11 inches in length and works well with small specimens such as squids, individual organs, and grass frogs. The large size is about 13 inches long and works well with specimens such as medium pigs, pigeons, and small dogfish. The extra-large size is approximately 30 inches long. It’s ideal for large dogfish, fetal horses, organ plucks, and similarly large specimens.
Accessories information: Dissecting pan set parts are available separately as well.
Product Family Options
Sets
Información del producto
  • Size
    Width
    Height
    Length
    Availability
    Price
  • 470201-230
    Pan With Pad
    Pan With Pad
    Standard
    19 cm
    4 cm
    29 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $434.93
    $434.93
    Each (1UN)
       $434.93
    Supplier Number: AS2
Pan
Información del producto
  • Size
    Width
    Height
    Length
    Availability
    Price
  • 470177-936
    Large Pan
    Large Pan
    Large
    24 cm
    4 cm
    33 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $315.53
    $315.53
    Each (1UN)
       $315.53
    Supplier Number: 148510
Covers
Información del producto
  • Size
    Width
    Height
    Length
    Availability
    Price
  • 470006-168
    Standard Cover
    Standard Cover
    Standard
    19 cm
    10 cm
    28 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $239.87
    $239.87
    Each (1UN)
       $239.87
    Supplier Number: 183646
  • 470006-172
    Large Cover
    Large Cover
    Large
    24 cm
    10 cm
    33 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $239.87
    $239.87
    Each (1UN)
       $239.87
    Supplier Number: 183647
Pads
Información del producto
  • Size
    Width
    Height
    Length
    Availability
    Price
  • 470006-166
    Standard Pad
    Standard Pad
    Standard
    19 cm
    2 cm
    29 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $197.22
    $197.22
    Each (1UN)
       $197.22
    Supplier Number: ASPAD
  • 470006-170
    Large Pad
    Large Pad
    Large
    24 cm
    2 cm
    33 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $293.14
    $293.14
    Each (1UN)
       $293.14
    Supplier Number: ADPAD