Large Overflow Can
Large Overflow Can
Catalog # 470110-882
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: OFLCN1
Jump to:
Product Details & Documents
For measuring the densities of larger objects.
- Dimensions are 10cm diameter by 13cm high
- The 3.5cm spout is 1.7cm below the lip, giving a capacity of about 850ml
A seamless drawn aluminum can with a rolled lip for extra strength.
Especificaciones
- Description:Large Overflow Can