Langenbeck Bone Saw
Langenbeck Bone Saw, Stainless steel, 110 mm (4.5")
470018-932
: 140972
When dissecting larger animals, saws will cut through tough bone much easier than shears. Long lasting stainless steel.
- Description:Langenbeck Bone Saw
- Length:110 mm (4.5")
- Material:Stainless steel