Lampboard
3- Lamp Lamp board
Catalog # 470014-932
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: LMPB03
Jump to:
Product Details & Documents
Study series and parallel circuits with this high quality apparatus.
- Includes lamps and an activity manual.
- 3 bulbs included
- Recommended bulb size is 1.5V and 3.8V (0.3 amps)
The connecting strips allow students to switch between series and parallel circuits. Comes on an acrylic base. Measures 5" x 10"
Especificaciones
- Description:3- Lamp Lamp board