United Scientific™ Lamp Sockets
Proveedor: UNITED SCIENTIFIC SUPPLIES
Certificados
Acerca de este artículo
Minature sockets for small bulbs.
Two terminals for easy conections
- All-metal construction
- Fits E10 screw base mini bulbs
These miniature metal sockets have two terminals and are compatible with all of our miniature screw base incandescent lamps. Package of 15.
Frequently Bought Together