Orders LinkContactUsLinkComponent
LaMotte® Soil pH TesTabs Kit
LaMotte® Soil pH TesTabs Kit
LaMotte Soil pH TesTabs Kit, 50
Catalog # 470021-532
Número de proveedor #:  5912
LaMotte® Soil pH TesTabs Kit
Catalog # 470021-532
CAS Number:  
LaMotte® Soil pH TesTabs Kit
LaMotte® Soil pH TesTabs Kit
LaMotte Soil pH TesTabs Kit, 50
CAS Number:  
Price
$703.57
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An Ideal Alternative to Liquid pH Indicators
  • No hazardous compounds required for testing
  • Colorimetric pH determination
  • Includes instructions
Quickly and simply measure soil pH with these non-hazardous reagents. The kit includes 50 tests and a laminated color chart for comparisons and pH determinations. Chart values feature range 4.0 to 11.0 pH. Distilled or deionized water recommended. Includes instructions.
Documents
Especificaciones
Especificaciones
  • Description:
    LaMotte Soil pH TesTabs Kit
  • No. of Tests:
    50
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 