LaMotte® pH TRACER
Acerca de este artículo
Store Up to 15 Readings
- Automatic Temperature Compensation
- Auto-off to Conserve Battery Life
- Rugged Flat Electrode
For clear readings everytime, this pH meter features a large, bold display, which shows pH and temperature of a sample in numbers or bar graphs. A built-in memory stores up to 15 readings. Automatic shut-off and low battery indicator are additional features. It includes 4, 7, and 10 pH buffer tablets and four LR-44 batteries. pH range: 0–14, resolution: 0.01 pH, accuracy: ±0.01 pH. Temperature range: 32–149°F (0–65°C), resolution: 0.1°F/C.
pH Range: 0 - 14. Resolution: 0.01 Accuracy: ±0.01 pH.
Temperature Range: 32–149 °F; 0–65 °C. Resolution: 0.1°F/°C.
2 Options Available Below
Product Details & Documents
Store Up to 15 Readings
- Automatic Temperature Compensation
- Auto-off to Conserve Battery Life
- Rugged Flat Electrode
For clear readings everytime, this pH meter features a large, bold display, which shows pH and temperature of a sample in numbers or bar graphs. A built-in memory stores up to 15 readings. Automatic shut-off and low battery indicator are additional features. It includes 4, 7, and 10 pH buffer tablets and four LR-44 batteries. pH range: 0–14, resolution: 0.01 pH, accuracy: ±0.01 pH. Temperature range: 32–149°F (0–65°C), resolution: 0.1°F/C.
pH Range: 0 - 14. Resolution: 0.01 Accuracy: ±0.01 pH.
Temperature Range: 32–149 °F; 0–65 °C. Resolution: 0.1°F/°C.
Specifications for Product Family
Especificaciones
- Catalog No:
- 470012-552
- 470012-554
- pH range:
- 0 - 14
- pH resolution:
- 0.01
- pH accuracy:
- ±0.01
- Temperature range:
- 32–149 °F; 0–65 °C
- Temperature resolution:
- 0.1°F/°C
Especificaciones
Product Family Options
Información del producto
- AvailabilityPrice
- 470012-552LaMotte® pH TRACERSupplier Number: 1741Especificaciones:
More
Specifications
Cat. No.470012-552pH range0 - 14pH resolution0.01pH accuracy±0.01Temperature range32–149 °F; 0–65 °CTemperature resolution0.1°F/°C - 470012-554Replacement Sensor for LaMotte® pH TRACERSupplier Number: 1733Especificaciones:
More
Specifications
Cat. No.470012-554pH rangepH resolutionpH accuracyTemperature rangeTemperature resolution