LaMotte® GREEN Low Cost Estuary Kit
Catálogo # 470146-188
Proveedor: LA MOTTE CHEMICAL PROD CO SE
Especificaciones
- Descripción:Kit, GREEN Low Cost Estuary, LaMotte Chemicals
- Referencia:470146-188
- Nº de proveedor:5911-20
Especificaciones
Acerca de este artículo
Designed to provide a simple, affordable, and nonhazardous method of testing water for nine basic water quality parameters, this kit contains everything a science class needs to test water samples.
- Step-By-Step Instructions Included
- Tests Include Coliform Bacteria, Salinity, and More
- Ten Tests of Each Parameter Except Coliform
- Nine Basic Water-Quality-Parameter Tests
The kit tests for coliform bacteria, salinity, dissolved oxygen, biochemical oxygen demand, nitrate, pH, phosphate, temperature, and turbidity.
Ten tests of each parameter are provided, except for coliform, which is supplied in two tests. The kit includes step-by-step instructions.