Orders LinkContactUsLinkComponent
LaMotte® Acidity Test Kit
LaMotte® Acidity Test Kit
Soil Acidity Test Kit, 4 - 8 pH
Catalog # 470201-278
Número de proveedor #:  5024
LaMotte® Acidity Test Kit
Catalog # 470201-278
CAS Number:  
LaMotte® Acidity Test Kit
LaMotte® Acidity Test Kit
Soil Acidity Test Kit, 4 - 8 pH
CAS Number:  
Price
$831.50
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Accurate pH Testing
  • 100 Tests
  • Spot Testing of Your Soil
Tests are made with a wide range indicator and give the pH of soil to ±0.5 pH units. Includes 50 tests, chemical indicator, porcelain plates, color comparator chart, complete Instructions, pocket-size case and a soil test handbook.
Especificaciones
Especificaciones
  • Description:
    Soil Acidity Test Kit
  • Range:
    4 - 8 pH
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 